missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Click to view available options
Gel Size:
12 x 14 cm
Unit Size:
Each
Description
Provide uniform, reproducible results with the Thermo Scientific™ Owl™ EasyCast™ B3 Gel System with Built-in Recirculation, ideal for long runs, multiple sample sets or RNA gels.
Features:
- Prevents the formation of pH and ionic gradients
- Delivers clear results for samples run over long time periods
- Requires no external pumps, tubing or stir bars, as recirculation system is built into buffer chamber
- Eliminates uneven migration, band distortion or disassociation of pH-dependent glyoxylated RNA molecules that can result when ionic depletion occurs
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast™ gasketed UVT gel tray
- 12- and 20-well, double-sided 1.0/1.5mm thick combs
Specifications
Specifications
| Certifications/Compliance | CE marked |
| Description | EasyCast; Gel Size: 12 x 14cmL; Buffer volume: 1L; L x W x H: 25 x 18 x 12cm |
| Gel Size | 12 x 14 cm |
| Product Line | Owl |
| Type | Mini Gel Electrophoresis System |
| Sample Size | 8 to 50 |
| Volume (Metric) Buffer | 1000 mL |
| Dimensions (L x W x H) | 9.75 x 7 x 4.72 in. (25 x 18 x 12 cm) |
| Length (English) | 9.75 in. |
| Length (Metric) | 25 cm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction