Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
131.00 € - 157.00 €
Specifications
Product Type Gel Drying Kit
Quantity Each
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12454637
View Documents
Thermo Scientific™
GDF-10
Gel-Drying Kit, Minigel Format This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11560435
View Documents
Thermo Scientific™
GDF-20
Gel-Drying Kit, Large Format
157.00 €
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
Description
Description
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
Features:
  • Available in minigel and large format sizes
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
  • Two sets of gel-drying frames
  • 50 sheets of precut cellophane
  • Two solid back plates
  • Eight side clips
Note:
  • Frames and cellophane are available separately
Specifications
Specifications
Gel Drying Kit