missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems

Product Code. p-4528751
Click to view available options
Dimensions (L x W x H):
25 x 25 x 5 cm
50.4 x 39.7 x 5.1 cm
Capacity:
Transfer area: 20 x 20 cm
Transfer area: 35 x 45 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12064927

Brand: Thermo Scientific™ HEP1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.

  • Fast, efficient transfer of proteins and nucleic acids to membranes
  • Buffer contained within blotting paper is sufficient for efficient transfer of molecules
  • Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate
  • Ideal for proteins less than 150 kDa

Includes:

  • Base and lid
  • Power supply leads
  • Heavy-duty knobs
TRUSTED_SUSTAINABILITY

Specifications

Capacity Transfer area: 20 x 20 cm
Product Line Owl
Type Electroblotting System
Dimensions (L x W x H) 25 x 25 x 5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.