missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ P1-CST Multiple Gradient Gel Caster, 10 teeth
Cast one to five gels simultaneously with Fisherbrand™ P1-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
21.00 € - 332.00 €
Product Code | Brand | Description | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Thickness (Metric) | Price | Quantity & Availability | ||||
12404677
|
Thermo Scientific™
P1-CST |
Gel Caster | 18 cm |
332.00 €
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||||
11987515
|
Thermo Scientific™
P1-SC |
Spacer (2 sides,1 bottom); 1.0cm x 0.8mm | 0.8 mm |
21.00 €
Pack of 2 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
Description
Cast one to five gels simultaneously with Fisherbrand™ P1-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab