Learn More
Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.
Brand:  Thermo Scientific™ P7-10FR
 View more versions of this product
Product Code. 12404697
  • 45.30 € / Each
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Frosted notched glass
Owl
0.09 in.
10 cm
Owl P7-CST System
Notched Glass
10 cm