Learn More
Thermo Scientific™ Owl™ S3S System Combs
Eliminate comb assembly with these heavy-duty, one-piece combs.
55.16 € - 67.85 €
Specifications
For Use With (Equipment) Owl™ S3S Aluminum-Backed Sequencer System
Product Type Combs
View More Specs
Products 4
Product Code Brand Description Teeth Count Thickness (Metric) Teeth Volume (Metric) Well Price Quantity & Availability  
Product Code Brand Description Teeth Count Thickness (Metric) Teeth Volume (Metric) Well Price Quantity & Availability  
12444717
View Documents
Thermo Scientific™
S2S-MT2
Sharktooth Micro-Well Comb 72 (2X) 0.4 mm 10µL
65.80 €
Each
12454717
View Documents
Thermo Scientific™
S2S-MT3
Sharktooth Comb 110 (3X) 0.4 mm 5.5µL
67.85 €
Each
11505195
View Documents
Thermo Scientific™
S2S-MT42
Sharktooth Micro-Well Comb 146 (4X) 0.2 mm 2µL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12474717
View Documents
Thermo Scientific™
S2S-MT44
Sharktooth Micro-Well Comb 146 (4X) 0.4 mm 3.7µL
65.80 €
Each
Description
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ S3S System Combs. These heavy-duty, one-piece combs are for use with the Owl™ S3S System.
Heavy duty, one piece combs eliminate comb assembly.
Specifications
Specifications
Owl™ S3S Aluminum-Backed Sequencer System
Combs