missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts.

14.70 € - 40.60 €

Specifications

For Use With (Equipment) Owl™ S4S Aluminum-Backed Sequencer System
View More Specs

Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11530495
View Documents
Thermo Scientific™
R10384
Buffer Drain Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12404737
View Documents
Thermo Scientific™
S4S-GK
Replacement Gasket
14.70 €
Each
Please to purchase this item. Need a web account? Register with us today!
11585235
View Documents
Thermo Scientific™
STK-10
Stopcocks (1 pair) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12454737
View Documents
Thermo Scientific™
TEMP-5
Adhesive temperature indicators (pkg. of 5)
40.60 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.

Choose from these replacement parts for optimal manual sequencing.
Specifications

Specifications

Owl™ S4S Aluminum-Backed Sequencer System
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.