missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Monoclonal Antibody (443318), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523676
This item is not returnable.
View return policy
Description
In Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFAS, rhGITR, rhHVEM, rhLT beta R, rhNGF R, rhRANK, rhRELT, rhTAJ, rhTNF RI, rhTNF RII, rhTRAIL, rhTRAIL R4, rhTRAIL R3, rhTWEAK R, or rhXEDAR is observed.
OX40 Monoclonal antibody specifically detects OX40 in Human samples. It is validated for Flow Cytometry.Specifications
OX40 | |
Monoclonal | |
PE | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Rat | |
Protein A/G | |
RUO | |
7293 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
443318 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P43489 | |
Tnfrsf4 | |
Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4 Leu29-Ala216 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction