missing translation for 'onlineSavingsMsg'
Learn More

Oxalacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11367138
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-42-7
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
MDL Number MFCD00002592
InChI Key KHPXUQMNIQBQEV-UHFFFAOYSA-N
Synonym oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
PubChem CID 970
ChEBI CHEBI:30744
IUPAC Name 2-oxobutanedioic acid
SMILES C(C(=O)C(=O)O)C(=O)O

Specifications

Melting Point 161°C (decomposition)
Flash Point 88°C (190°F)
Assay Percent Range >98%
Quantity 2 g
UN Number UN3261
Beilstein 1705475
Merck Index 14,6909
Solubility Information Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate.
Formula Weight 132.07
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Oxalacetic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.