Learn More
Oxalate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
54.00 € - 114.00 €
Chemical Identifiers
Molecular Formula C2O42
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12922175
View Documents
Fisher Chemical
A 035 100
100 mL
54.00 €
100mL
12932175
View Documents
Fisher Chemical
A 035 250
250 mL
77.00 €
250mL
12942175
View Documents
Fisher Chemical
A 035 500
500 mL
114.00 €
500mL
Specifications
Chemical Identifiers
C2O42
Specifications
1g/L
Single-component Standard Solution
H2O
Oxalate
Bottle
C2O42
Liquid
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)