missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 15491347
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Oxalic acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144-62-7
Quantity 1 L
Molecular Formula C2H2O4
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.034
PubChem CID 971
ChEBI CHEBI:16995
Formula Weight 90.04
Solubility Miscible with water, alcohol and ether
Physical Form Liquid
Chemical Name or Material Oxalic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'