missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, ACS, 99.5-102.5%, Thermo Scientific Chemicals

20.90 € - 444.21 €

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387568
View Documents
Thermo Scientific Alfa Aesar
033262.18
50 g
20.90 €
50g
Please to purchase this item. Need a web account? Register with us today!
11317578
View Documents
Thermo Scientific Alfa Aesar
033262.30
250 g
61.31 €
250g
Please to purchase this item. Need a web account? Register with us today!
11397568
View Documents
Thermo Scientific Alfa Aesar
033262.A1
1 kg
126.63 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11307578
View Documents
Thermo Scientific Alfa Aesar
033262.A7
5 kg
444.21 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing and as a reagent for dehydration and condensation Oxalic acid dihydrate acts as an analytical reagent, as a reducing agent for photographic films and as mordant in the dyeing process. It is used as a plating agent, a surface treating agent, a grinding agent, a precipitating agent in rare-earth mineral processing, a bleaching agent in textile and a purifying agent in the pharmaceutical industry. It plays an important role in antibiotic medication like oxytetracycline and chloramphenicol. It finds application in water treatment plants to remove calcium waste water. It is also used as a buffer in chromatographic separation as well as bleach for wood, removing black stains caused by water penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation
Specifications

Chemical Identifiers

6153-56-6
126.064
GEVPUGOOGXGPIO-UHFFFAOYSA-N
61373
C(=O)(C(=O)O)O.O.O
C2H6O6
MFCD00149102
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
oxalic acid;dihydrate

Specifications

6153-56-6
99.5-102.5%
HO2CCO2H·2H2O
UN3261
14,6911
Soluble in water,ethanol,ether and glycerol; Insoluble in chloroform.
C(=O)(C(=O)O)O.O.O
126.064
126.07 (90.04 Anhydrous)
ACS
Crystalline
101°C to 105°C
C2H6O6
MFCD00149102
3679436
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid;dihydrate
61373
99.5 to 102.5%
1.65
Oxalic acid dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312

P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P363-P501c

H302+H312-H318-H373

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 205-634-3

missing translation for 'rtecsNumber' : KI1600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Oxalic acid dihydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Oxalic acid dihydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.