missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Oxalic Acid Dihydrate, Extra Pure, SLR

Specifications
Specifications
Packaging | Plastic powder jar |
Quantity | 1 kg |
Vapor Pressure | 21.5mbar 50 |
Physical Form | Powder/Solid |
Color | White |
pH | 1.3 |
Melting Point | 98°C |
Formula Weight | 126.04g/mol |
Safety and Handling


Product Identifier
- Oxalic acid dihydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oxalic Acid Dihydrate, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction