missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, TraceSELECT™, ≥99.9999% (Metals basis), Honeywell™ Fluka™

Product Code. 15657510 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15657510

Brand: Honeywell-Fluka 93722100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™,≥99.9999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

pH 1 (126.1g/L at 25°C)
CAS 6153-56-6
Melting Point 104°C to 106°C
Color Colorless
Molecular Formula C2H6O6
Linear Formula HO2CCO2H · 2H2O
MDL Number MFCD00149102
UN Number NONH for all modes of transport
Beilstein 3679436
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.O.O
IUPAC Name oxalic acid;dihydrate
Molecular Weight (g/mol) 126.064
PubChem CID 61373
Formula Weight 126.07g/mol
Percent Purity ≥99.9999% (metals basis)
Grade TraceSELECT
Quantity 100 g
Packaging Poly bottle
Physical Form Crystalline
Vapor Density 4.4 (vs air)
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.