missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, Thermo Scientific Chemicals

Artikelnummer. 10397190
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Spezifikation

Color Colorless
Linear Formula HO2CCO2H
Beilstein 02, 502
Merck Index 15, 7010
Solubility Information Solubility in water: soluble in water.
Concentration 0.0998 to 0.1002N (20°C)
Formula Weight 90.04
Density 0.9900g/mL
Quantity 1 L
Packaging Plastic bottle
Physical Form Liquid
Specific Gravity 0.99
Chemical Name or Material Oxalic acid
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'