missing translation for 'onlineSavingsMsg'
Learn More
Oxalyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 10234990

Oxalyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10234990
1 kg, Glass bottle
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10234990

Brand: Thermo Scientific Acros 129610010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-37-8, 75-44-5, 76-02-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.93
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl

Specifications

Melting Point -12.0°C
Color Colorless to Yellow
Density 1.4780g/mL
Boiling Point 63.0°C to 64.0°C (763.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCOCl
Refractive Index 1.4285 to 1.4305
Quantity 1 kg
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Merck Index 15, 7013
Specific Gravity 1.478
Solubility Information Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether
Formula Weight 126.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468321 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number129610025,129611000,129610250,129610010
Lot NumberA0465447 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number129610025,129611000,129610250,129610010
Lot NumberA0438812 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number129610025,129611000,129610250,129610010
Lot NumberA0450920 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number129610025,129611000,129610250,129610010
Lot NumberA0411678 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number129610025,129611000,129610250,129610010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxalyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.