missing translation for 'onlineSavingsMsg'
Learn More
Oxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals Product Code.: 15494028

Oxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15494028
250 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15494028

Brand: Thermo Scientific Alfa Aesar H57578.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Moisture sensitive. . Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114012-41-8
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD08669505
InChI Key UWOTZNQZPLAURK-UHFFFAOYSA-N
Synonym 3-oxetanecarboxylic acid, 3-oxetanecarboxylicacid, acmc-20ahnr, oxetane-3-carboxylc acd, oxetane-3-carboxylicacid
PubChem CID 19847174
IUPAC Name oxetane-3-carboxylic acid
SMILES C1C(CO1)C(=O)O

Specifications

Density 1.374
Boiling Point 244°C
Flash Point 115°C (239°F)
Quantity 250 mg
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 102.09
Percent Purity 95%
Chemical Name or Material Oxetane-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT18J004 Certificate TypeCertificate of Analysis Date26/07/2023 Catalog NumberH57578.ME,H57578.MD
Lot NumberS20J026 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberH57578.ME,H57578,H57578.MD
Lot NumberT08I024 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog NumberH57578.ME,H57578.MD
Lot NumberP26A002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57578.ME,H57578.MD
Lot NumberT03E027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57578.ME,H57578.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.