missing translation for 'onlineSavingsMsg'
Learn More

Oxibendazole, Thermo Scientific™

Product Code. 17525555
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 20559-55-1
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number 00133728
InChI Key RAOCRURYZCVHMG-UHFFFAOYSA-N
IUPAC Name methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate
SMILES CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Packaging Glass Bottle
Quantity 5 g
Color White to Beige
Certificate of Analysis Available
Merck Index 13-7005
Solubility Insoluble in water
Physical Form Powder
Chemical Name or Material Oxibendazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.