Learn More
Oxidation Reagent 0.05 M Iodine in 90/10 Pyridine/Water (v/v), BioSyn™, Honeywell Burdick & Jackson™
BioSyn™ Honeywell Burdick & Jackson™ reagent solvents for DNA/RNA synthesis
Brand:  Honeywell Burdick & Jackson BR664-4X2.5GL
Product Code. 16885421
  • 1043.86 € / Pack of 4
Explore more special offers
Description
Description
  • Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of application
  • All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oxidation reagent
7553-56-2
0.013
4 x 2.5 L
Clear liquid
−42.2°C
Colorless to yellow
115.25°C (1.013 hPa)
GL45 Glass Bottle
BioSyn™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Boiling and melting point temperatures are based off the physical main component.