missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Burdick & Jackson™ Oxidation Reagent, 0.05M Iodine in 90/10 Pyridine/Water (v/v)

BioSyn™, for DNA/RNA synthesis

267.00 €

Specifications

CAS 7553-56-2
Density 0.983 g/cm3
View More Specs

Products 1
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
16875421
View Documents
Honeywell Burdick & Jackson™
BR664-2.5GL
4 x 2.5 L GLASS BOTTLE, GL45 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications

Specifications

7553-56-2
0.983 g/cm3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.