Learn More
Oxidation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 70/20/10 THF/Pyridine/Water (v/v)
Brand:  Honeywell Fluka BR666-4X2.5L
 View more versions of this product
Product Code. 16805431
  • 1296.80 € / Pack of 4
Explore more special offers
Description
Description
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7553-56-2, 109-99-9, 7732-18-5, 110-86-1
66°C
-15°C
Liquid, clear
0.50%, 67.20%, 10.90%, 21.40%
4 x 2.5 L
Glass Bottle
Oxidation reagents