Learn More
Oxidation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 78/20/2 THF/Pyridine/Water (v/v)
Brand:  Honeywell Fluka BR663-4X2.5L
Product Code. 16865421
  • 1433.96 € / Pack of 4
Explore more special offers
Description
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
109-99-9, 110-86-1, 7732-18-5, 7553-56-2
66°C
-15°C
Liquid, clear
78.00%, 20.00%, 2.00%, 0.20%
4 x 2.5 L
Glass Bottle
Oxidation reagents