Learn More
WTW™ OxiTop™ IS BOD Self Check Measurement System
Click to view available options
Type:
BOD Meter, 2 Heads
Tamaño de la unidad:
Pack of 2
Descripción
- Easy handling without dilution series
- Data security by built-in measured value memory
- Suitable for immediate use
- Based on pressure management in a closed system - the microorganisms in the sample consume the oxygen and generate CO&sub2; in the process
- OxiTop™ heads have an AutoTemp function
- Automatic saving of 5 measured values, manual reading of values after 5 days have elapsed
Especificaciones
Especificaciones
| Type | BOD Meter, 2 Heads |
| Display Type | LED |
| Range | 0 to 4000 mg/L BSB |
| For Use With (Application) | Respirometric self-check measurement |
| Includes | 2 OxiTop measuring heads (yellow, green), 2 PF600 sample bottles, 2 GK 600 Rubber sleeves and 2 RST 600 magnetic stirrers |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido