Learn More
WTW™ OxiTop™ IS BOD Self Check Measurement System
A complete respirometric measuring system for BOD self-check with undiluted samples featuring ranges of up to 4,000mg/L BOD.
Brand: WTW™ 208817
This item is not returnable.
View return policy
Description
- Easy handling without dilution series
- Data security by built-in measured value memory
- Suitable for immediate use
- Based on pressure management in a closed system - the microorganisms in the sample consume the oxygen and generate CO&sub2; in the process
- OxiTop™ heads have an AutoTemp function
- Automatic saving of 5 measured values, manual reading of values after 5 days have elapsed
Specifications
OxiTop™ Set-6 Manual Measuring Unit | |
0 to 4000 mg/L BSB | |
6 OxiTop™ measuring heads (3 yellow, 3 green), 6 PF600 sample bottles, 6 GK 600 Rubber sleeves and 6 RST 600 magnetic stirrers |
LED | |
Respirometric self-check measurement |