missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Oxivir Excel Concentrate

Product Code. p-7221342
Click to view available options
Description:
For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 17231614

Brand: Diversey™ 100941436

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly concentrated one step cleaner disinfectant for non-invasive medical devices and nonporous surfaces.

  • Effective against Bacteria, Viruses and Yeasts
  • Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
  • Extensively tested against European norms
  • Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
  • Excellent user safety profile reduces worker risk
  • No hazard warnings in dilution
  • Based on Diversey's patented AHP formulation (Accelerated Hydrogen Peroxide)
  • AHP formulation is gentle on surfaces extending equipment life
  • Does not contain respiratory irritants
  • Medical Device Class IIa
  • Oxivir Excel can be used on any hard surface usually encountered in a laboratory, production and non-production environments including stainless steel, glass, acrylics and PVC. It is not suitable for use on marble, copper, brass or other surfaces sensitive to oxidative agents.

Specifications

Product Type Disinfectant Concentrate
pH 1.95
Sterility Non-sterile
Composition Patented Accelerated Hydrogen Peroxide formulation (conc.)
Container Type Jug
Description For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
Quantity 2 Pk.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • Oxivir Excel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Supplemental information
  • MIXTURE LIST-Contains : alkylbenzenesulphonic acid, propanol, hydrogen peroxide

The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.