missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Agar Bacteriological
Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.
341.00 € - 2270.00 €
Specifications
Description | Agar No. 1, bacteriological |
---|---|
Form | Powder |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10351303
|
Thermo Scientific™
LP0011B |
500 g |
341.00 €
500g |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
10055782
|
Thermo Scientific™
LP0011T |
5 kg |
2270.00 €
5kg |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.
Specifications
Agar No. 1, bacteriological | |
Powder | |
Agar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title