Learn More
Thermo Scientific™ Oxoid™ Anaerobic Low Temperature Catalyst
Oxoid Anaerobic Low Temperature Catalyst is a safety catalyst designed to stay cooler than traditional catalysts.
Brand: Thermo Scientific™ BR0042A
This item is not returnable.
View return policy
Description
Oxoid Anaerobic Low Temperature Catalyst is a safety catalyst designed to stay cooler than traditional catalysts.
- To be changed every 30 cycles
Specifications
Anaerobic Low Temperature Catalyst | |
Each |