missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker Agar

Easily isolate and enumerate Staphylococcus aureus with highly selective and sensitive, ready-to-use media.

30.55 €

Specifications

Format 90mm Monoplate
Media Type Baird-Parker Agar
Product Type Agar
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
View More Specs

Products 1
Product Code Brand Quantity Media Type Fill Weight Manufacturing Origin Price Quantity & Availability  
Product Code Brand Quantity Media Type Fill Weight Manufacturing Origin Price Quantity & Availability  
12972588
View Documents
Thermo Scientific™
PO0168A
10/Pk. Baird-Parker Agar 19.5g ± 1g United Kingdom
30.55 €
Pack of 10
Estimated Shipment: 27-12-2024
to see stock.
Edge
Add to basket
 
Description

Description

Isolate and enumerate Staphylococcus aureus with ready-to-use, highly selective and sensitive Thermo Scientific™ Baird-Parker Agar. The medium aids the recovery of damaged cells for presumptive identification of Staphylococcus aureus in food specimens.

Baird-Parker Agar medium is widely recommended by national and international bodies (ISO 6888-1) for the isolation of Staphylococcus aureus1.

  • Staphylococcus aureus forms distinctive grey-black shiny colonies with surrounding clear zones
  • Ready-to-use convenience of prepared media
  • The formulation conforms to ISO 6888-1
Baird-Parker2 developed this medium from the tellurite-glycine formulation of Zebovitz et al.3 and improved its reliability in isolating Staphylococcus aureus from foods. The addition of sodium pyruvate protects damaged cells and aids their recovery4 and egg yolk emulsion acts as a diagnostic agent.

The selective agents, glycine, lithium and tellurite, have been carefully balanced to suppress the growth of most bacteria present in foods, without inhibiting Staphylococcus aureus.

Egg yolk emulsion makes the medium yellow and opaque. Staphylococcus aureus reduces tellurite to form grey-black shiny colonies and then produces clear zones around the colonies by proteolytic action. On further incubation, most strains of Staphylococcus aureus form opaque haloes around the colonies.

Some strains of Staphylococcus saprophyticus may also produce both clear zones and opaque haloes but can be distinguished by the longer incubation time required5.

Colonies typical of Staphylococcus aureus, but without an egg yolk reaction, should also be tested for coagulase production6. Egg yolk reaction negative strains of Staphylococcus aureus may occur in some foods, especially cheese.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery, we recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications

Specifications

90mm Monoplate
Agar
10/Pk.
Baird-Parker Agar
Wrapped in single cellulose-based film wrap
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 Weeks

SDS
Documents

Documents

Product Certifications