missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Biochemical Identification System (O.B.I.S.) Salmonella

Rapidly differentiate Salmonella spp. from other organisms exhibiting similar colonial appearance with this rapid colorimetric test.

Brand:  Thermo Scientific™ ID0570M

Product Code. 11473969

  • 294.00 € / 60 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Salmonella Antigen
O.B.I.S. Salmonella test cards, O.B.I.S. PYR developing solution, O.B.I.S. buffer solution, instruction leaflet
Salmonella
O.B.I.S. Salmonella
Salmonella Test Kit
60 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6146304 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog NumberID0570M
Lot Number6133220 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog NumberID0570M
Lot Number3734414 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberID0570M
Lot Number3639504 Certificate TypeCertificate of Analysis Date23/03/2023 Catalog NumberID0570M
Lot Number3531338 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog NumberID0570M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Biochemical Identification System (O.B.I.S.) Salmonella >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.