missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ C-F-C Supplement

Oxoid C-F-C Supplement is used for the isolation of Pseudomonas species.

Brand:  Thermo Scientific™ SR0103E

51.37 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 11434179

  • 88.30 € / Pack of 10
Estimated Shipment: 27-11-2024
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Oxoid C-F-C Supplement is used for the isolation of Pseudomonas species.

  • Add to to Pseudomonas Agar Base, Part No. CM0559B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C-F-C Supplement
Microbiology Media
Each vial for 500mL medium
Freeze-dried
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers