missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Ceftazidime Antimicrobial Susceptibility discs

Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.

Brand:  Thermo Scientific™ CT1629B

 View more versions of this product

Product Code. 10481303

  • 67.20 € / Pack of 250

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Ceftazidime Antimicrobial Susceptibility discs.

Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.

disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ceftazidime
10μg
CAZ
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with DIN, EUCAST, SFM guidelines
Oxoid Ceftazidime Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224276 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog NumberCT1629B
Lot Number6242014 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberCT1629B
Lot Number6230146 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberCT1629B
Lot Number6224277 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberCT1629B
Lot Number6196849 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberCT1629B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Ceftazidime Antimicrobial Susceptibility discs >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.