missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Chloramphenicol Selective Supplement
Oxoid Chloramphenicol Selective Supplement assists in the isolation of yeast and molds associated with spoilage.
49.90 € - 129.00 €
Specifications
Description | Chloramphenicol Selective Supplement |
---|---|
Form | Freeze-dried |
Product Type | Selective Supplement |
Quantity | 10/Pk. |
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
11464159
|
Thermo Scientific™
SR0078E |
Each vial for 500mL medium |
49.90 €
Pack of 10 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
12771860
|
Thermo Scientific™
SR0078H |
Each vial for 2L medium |
129.00 €
Pack of 10 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
For the isolation of yeast and molds associated with spoilage, add Oxoid Chloramphenicol Selective Supplement to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.
For the isolation of yeast and molds associated with spoilage, add to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.Specifications
Chloramphenicol Selective Supplement | |
Freeze-dried | |
Selective Supplement | |
10/Pk. |