missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromogenic Cronobacter Isolation Agar (Dehydrated)

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with this chromogenic medium.

Brand:  Thermo Scientific™ CM1122B

Product Code. 13215929

  • 1050.00 € / 500g
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with Thermo Scientific™ Oxoid™ Chromogenic Cronobacter Isolation Agar (Dehydrated). Chromogenic Cronobacter Isolation Agar (CCI) is an enhanced formulation of BrillianceCronobacter Sakazakii Agar (DFI) with improved specificity and colony coloration for the detection of Cronobacter.

Cronobacter sakazakii is a Gram-negative rod-shaped bacterium that rarely causes disease in healthy adults. It has been implicated in outbreaks of disease in premature infants (neonates). Research suggests that neonates, or those infants who have other medical conditions, are more susceptible to this infection. Most reported cases of infection are severe, including sepsis (bacteria in the blood, meningitis, or necrotizing enterocolitis (severe intestinal infection).

Use Chromogenic Cronobacter Isolation Agar for differentiation and enumeration of Cronobacter sakazakii from infant formula and other food samples.

  • Selective: due to the presence of desoxycholate the medium is selective against most Gram-positive organisms.
  • Easy-to-read: Cronobacter sakazakii produces blue-green colonies on this medium.

Cronobacter spp. have been isolated at low levels from powdered infant formulae, and the organisms’ high tolerance to desiccation provides a competitive advantage in the dry environments of milk powder factories, increasing the risk of product contamination1.

Chromogenic Cronobacter Isolation (CCI) agar is an optimisation of the original Brilliance™ Enterobacter Sakazakii Agar (DFI), developed by Iversen et al. in 20043. The concentration of the X-α-glucoside chromogen was increased to improve the blue-green colouration of Cronobacter strains with weak glucosidase activity. Sodium thiosulphate and ammonium iron (III) citrate are included to differentiate hydrogen sulphide-producing Enterobacteriaceae, which are coloured grey/brown, and sodium deoxycholate is added to inhibit the growth of Gram-positive competitors.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chromogenic Cronobacter Isolation Agar
Agar
For 16.3L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only