missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Burkholderia cepacia Medium

Isolate Burkholderia cepacia from patients with cystic fibrosis and from non-sterile inorganic salt solutions used in hospitals.

Brand:  Thermo Scientific™ CM0995B

Product Code. 10026402

  • 226.00 € / 500g
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate Burkholderia cepacia from respiratory secretions of patients with cystic fibrosis with Thermo Scientific™ Oxoid™ Burkholderia Cepacia Agar Base, which is also used for routine testing of non-sterile inorganic salt solutions in hospital environments. The slower growing B. cepacia can be missed on conventional media, such as blood or MacConkey Agar, due to overgrowth caused by other faster growing organisms found in the respiratory tract of CF patients, such as mucoid Klebsiella species, Pseudomonas aeruginosa and Staphylococcus species.This may lead to the infection being missed or wrongly diagnosed.

Burkholderia cepacia (formerly known as Pseudomonas cepacia) is a motile aerobic oxidase positive Gram-negative bacillus commonly found in liquid reservoirs and moist environments. The cells are 0.5 to 1.0mm wide and 5mm in length. It is an important opportunistic pathogen and causes pulmonary infection among individuals with cystic fibrosis (CF). Isolates from CF patients often display multidrug resistance and as many as 20% of colonised individuals will succumb to Burkholderia cepacia syndrome, a necrotizing pneumonia associated with fever that culminates into a rapid and fatal clinical deterioration1.

Burkholderia Cepacia Agar Base is based on PC Medium originally devised by Gilligan et al. where it was shown to be superior for growth of Burkholderia cepacia after 48 hours when compared to MacConkey2. Typical Burkholderia cepacia colonies are circular and sage green, the medium turning from straw-green to bright pink during incubation. All colonies should be further identified and confirmed.

Originally isolated from onions, Burkholderia cepacia can survive for long periods and multiply in hostile environments, such as antiseptic and disinfectant solutions, distilled water, whirlpool baths, nebulizers and commercially packaged urinary catheter kits3. An outbreak in Arizona in 1998 due to contaminated alcohol-free mouthwash was investigated by the Food and Drug Administration (FDA), who suggested an association with the deionization procedure of the water used to prepare the product4. The organism may be present in low numbers in many non-sterile products used in hospitals. It has been isolated from various water sources and can grow in distilled water with a nitrogen source due to its ability to fix carbon dioxide from air5. Suction catheters rinsed in acetic acid solution have reduced incidence of transmission of Burkholderia cepacia and other pseudomonads.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Burkholderia Cepacia Agar Base
Agar
For 13.7L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For in vitro diagnostic use.