Learn More
Thermo Scientific™ M.R.S. Agar
Isolate and enumerate lactic acid bacteria from food samples with this medium.
Brand: Thermo Scientific™ CM0361B
121.06 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ M.R.S. Agar (Dehydrated). This medium is designed to support profuse growth of all strains of lactobacilli, even the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti.
M.R.S. Agar is designed to encourage the growth of the ‘lactic acid bacteria’ which includes species of the following genera: Lactobacillus, Streptococcus, Pediococcus, and Leuconostoc. All these species can produce lactic acid in considerable amounts.
- Superior performance over the tomato juice medium of Briggs1 and the meat extract tomato juice medium of de Man
- Allows growth of growth of all strains of lactobacilli, especially the difficult and slow-growing strains of Lactobacillus brevis and Lactobacillus fermenti
‘Lactic acid bacteria’ are Gram-positive, catalase and oxidase negative and are fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the ‘lactic acid bacteria’ show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Industrial Reference: AFNOR, IDF, NMKL standards. | |
Powder | |
500 g |
M.R.S. Agar (Dehydrated) | |
Agar | |
For 8.1L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only