missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement

Thermo ScientificTM OxoidTM ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.

Brand:  Thermo Scientific™ SR0242B

Product Code. 12986736

  • 232.00 € / Pack of 10
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.

  • Add to ONE Broth-Salmonella Part No. CM1091B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Salmonella species
Freeze-dried
10/Pk.
ONE Broth-Salmonella Supplement
Microbiology Media
Each vial for 225 mL mediμm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers