Learn More
Thermo Scientific™ Oxoid™ Peptone Bacteriological
The Oxoid Peptone Bacteriological is for use whenever a high quality bacteriological peptone is required.
192.00 €
Specifications
Description Peptone Bacteriological
Form Powder
Product Type Peptone
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10404013
View Documents Promotion Details
Thermo Scientific™
LP0037B
500 g
192.00 €
500g
Estimated Shipment: 21-11-2024
to see stock.
Add to basket
Description
Description
The Oxoid Peptone Bacteriological is for use whenever a high quality bacteriological peptone is required.
A nutritious, all-purpose peptone specially prepared for use with other refined culture media ingredients
Specifications
Specifications
Peptone Bacteriological
Powder
Peptone