Learn More
Thermo Scientific™ Oxoid™ Qualitative Eia
Brand: Thermo Scientific™ K604411-2
This item is not returnable.
View return policy
Description
IDEIA Norovirus, for the direct detection of Norovirus antigen in human faecal specimens, provides an accurate result within 2 hours of receiving a faecal specimen OxoidSafety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.