missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Liquid Medium EP/USP/JP/BP (Dehydrated)

Confidently perform sterility testing and/or determine the fungistatic activity of pharmaceutical products with this medium.

Brand:  Thermo Scientific™ CM0147B

Product Code. 10299472

  • 80.95 € / 500g
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Confidently perform sterility testing and/ or determine the fungistatic activity of pharmaceutical products with Thermo Scientific™ Oxoid™ Sabouraud Liquid Medium (Dehydrated). The medium is mycological sterility test medium conforming to the medium described in USP1 for the determination of fungistatic activity of pharmaceutical and cosmetic products in order to avoid false sterility tests.Sabouraud Liquid Medium also conforms to the parameters from the harmonised EP/USP/ JP Microbial Limit Testing for the microbial enumeration tests and tests for specified microorganisms2,3.

A suitable medium for the isolation of Candida albicans.

In clinical microbiology, the use of Sabouraud Dextrose Liquid Medium has been shown to increase the isolation rate of Candida albicans in blood culture4.

The medium may also be used for the cultivation of acidophilic bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sabouraud Liquid Medium
Dehydrated Culture Media
For 16.7L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only