Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
110.00 € - 117.00 €
Specifications
Description | Oxoid™ Vancomycin Supplement |
---|---|
Product Type | Microbiology Media |
Quantity | 10/Pk. |
Product Code | Brand | Format | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Quantity | Price | Quantity & Availability | ||||
12638797
|
Thermo Scientific™
SR0247E |
10/Pk. |
110.00 €
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
13285399
|
Thermo Scientific™
SR0186E |
10/Pk. |
117.00 €
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
- Each vial supplements 500mL of medium
Spécification
Oxoid™ Vancomycin Supplement | |
Microbiology Media | |
10/Pk. |