Learn More
Thermo Scientific™ WL Nutrient Agar (Dehydrated)
Isolate and enumerate microbial flora in brewing and fermentation processes with this medium.
Brand: Thermo Scientific™ CM0309B
108.16 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Isolate and enumerate microbial flora in brewing and fermentation applications with Thermo Scientific™ Oxoid™ WL Nutrient Agar (Dehydrated). The medium is based on that of Green and Gray1 and is suitable for the differentiation of wild yeasts from brewing yeasts2.
WL Nutrient Agar is recommended for the determination of the microbial flora in brewing and fermentation processes.
Reliable counts of fermentation process are obtained with the medium at 5.5. Adjustment to pH 6.5 facilitates the counting of bakers’ and distillers’ yeasts.
Cycloheximide (SR0222) may be added to the medium at a final concentration of 4 mg/L, to suppress yeast growth, the medium becomes selective for the bacterial contaminants of yeast cultures.
When making microbial counts with this medium the time and temperature of incubation will vary according to the materials tested and the organisms sought. Temperatures of 25°C are used for brewing materials and 30°C for bakers’ yeasts.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
WL Nutrient Agar | |
Agar | |
For 6.7L medium |
Powder | |
500 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only