Learn More
Thermo Scientific™ Oxoid™ X & V Factor Discs
For identification of Haemophilus spp.
Brand:  Thermo Scientific™ DD0005B
Product Code. 11499858
  • 51.65 € / Pack of 250
Explore more special offers
Description
Description
For identification of Haemophilus spp.
50 Disks/Cartridge
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Haemophilus species
V Factor Disc
5 x 50 Discs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6215241 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberDD0005B
Lot Number6199997 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberDD0005B
Lot Number6130673 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog NumberDD0005B
Lot Number6130674 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog NumberDD0005B
Lot Number3704935 Certificate TypeCertificate of Analysis Date06/01/2024 Catalog NumberDD0005B
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ X & V Factor Discs > 5 x 50 discs