Learn More
Hirschmann™ Oxygen Bottles
Individually marked and calibrated oxygen bottles acc. To winkler
Brand:  Hirschmann™ 6500187
Product Code. 11712293
  • 78.00 € / Pack of 2
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to Winkler
  • White graduation
  • Precisely adjusted
  • With labelling square
  • Consecutively serial-numbered on the bottle and the bevelled glass stopper
Specifications
Specifications
200 to 300 mL
135 mm