missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline hydrochloride, can be used as secondary standard'

Product Code. 10287220
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Type Oxytetracycline Hydrochloride
Physical Form Crystalline Powder
CAS 2058-46-0
Infrared Spectrum Authentic
Molecular Formula C22H25ClN2O9
MDL Number MFCD00135815
Beilstein 14, IV, 2633
Merck Index 15, 7075
Packaging Glass Bottle
Synonym 5-Hydroxytetracycline hydrochloride
Solubility Information Solubility in water: >100g/L. Other solubilities: 20g/L methanol
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 496.897
PubChem CID 121235537
Formula Weight 496.9
Percent Purity ≥97.0% Total Base (anhydrous and solvent free), ≥80 wt% (as is, calculated, HPLC)
Color Beige to Yellow
Quantity 500 mg
Chemical Name or Material Oxytetracycline hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'