Learn More
Macherey-Nagel™ Ozone Test Strips
Brand: Macherey-Nagel™ 90736
This item is not returnable.
View return policy
Specifications
White to Brown | |
Ozone testing | |
Ozone | |
Ozone |
Strip | |
<90, 90 to 150, 150 to 210, >210μg/m3 O3 | |
12 Pack | |
Test Strip |