Learn More
Welch™ P 6 Z Chemvac Combination Pump System
Brand: Welch™ 109030
This item is not returnable.
View return policy
Specifications
230 V 50/60 Hz | |
P 6 Z | |
50/60 Hz | |
500 mm | |
2 x 10-3 mbar |
330 mm | |
97/110 L/min. | |
340 mm | |
Chemvac Combination Pump System | |
230 V |