missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Product Code. 10554521
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2186-92-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00036507
InChI Key NNHYAHOTXLASEA-UHFFFAOYSA-N
Synonym 1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
PubChem CID 75140
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
SMILES COC(OC)C1=CC=C(OC)C=C1

Specifications

Color Colorless to Yellow
Density 1.0680g/mL
Boiling Point 235.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5045 to 1.5065
Quantity 25 g
Specific Gravity 1.068
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,soluble in common organic solvents
Formula Weight 182.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material p-Anisaldehyde dimethyl acetal
Show More Show Less
Product Identifier
  • p-Anisaldehyde dimethyl acetal

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'