missing translation for 'onlineSavingsMsg'
Learn More

p-Benzoquinone, 98+%, Thermo Scientific Chemicals

32.10 € - 621.00 €

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6H4O2
Molecular Weight (g/mol) 108.096
MDL Number MFCD00001591
InChI Key AZQWKYJCGOJGHM-UHFFFAOYSA-N
Synonym p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
PubChem CID 4650
ChEBI CHEBI:16509
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C=CC1=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11468690
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13162.22
100 g
32.10 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11454544
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13162.30
250 g
74.70 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11444544
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13162.0B
1000 g
148.00 €
1000g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11464544
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13162.0I
5000 g
621.00 €
5000g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p-Benzoquinone is used as a dienophile in Diels-Alder cycloadditions to prepare naphthoquinones and 1,4-phenanthrenediones. It acts as a dehydrogenation reagent and an oxidizer in synthetic organic chemistry. In the Thiele-Winter reaction, it is involved in the preparation of triacetate of hydroxyquinol by reacting with acetic anhydride and sulfuric acid. It is also used in the synthesis of bromadol and to suppress double- bond migration during olefin metathesis reactions. It is used as a precursor to hydroquinone which finds application in photography and as a reducing agent and an antioxidant in rubber production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Benzoquinone is used as a dienophile in Diels-Alder cycloadditions to prepare naphthoquinones and 1,4-phenanthrenediones. It acts as a dehydrogenation reagent and an oxidizer in synthetic organic chemistry. In the Thiele-Winter reaction, it is involved in the preparation of triacetate of hydroxyquinol by reacting with acetic anhydride and sulfuric acid. It is also used in the synthesis of bromadol and to suppress double- bond migration during olefin metathesis reactions. It is used as a precursor to hydroquinone which finds application in photography and as a reducing agent and an antioxidant in rubber production.

Solubility
Soluble in water, ethanol, ether, methanol, benzene, acetone and ethyl acetate.

Notes
Light sensitive. Store in cool place. Incompatible and reacts violently with strong oxidizing agents.
Specifications

Chemical Identifiers

106-51-4
108.096
AZQWKYJCGOJGHM-UHFFFAOYSA-N
4650
cyclohexa-2,5-diene-1,4-dione
C6H4O2
MFCD00001591
p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
CHEBI:16509
C1=CC(=O)C=CC1=O

Specifications

106-51-4
1.318
77°C (171°F)
Pungent
MFCD00001591
773967
14,8074
Soluble in water,ethanol,ether,methanol,benzene,acetone and ethyl acetate.
C1=CC(=O)C=CC1=O
108.096
CHEBI:16509
≥98%
112°C to 115°C
∼180°C (sublimation)
≥98%
C6H4O2
UN2587
Light sensitive
p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
AZQWKYJCGOJGHM-UHFFFAOYSA-N
cyclohexa-2,5-diene-1,4-dione
4650
108.1
p-Benzoquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c

H301+H331-H315-H319-H335-H500

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZOQUINONE

missing translation for 'einecsNumber' : 203-405-2

missing translation for 'rtecsNumber' : DK2625000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only