missing translation for 'onlineSavingsMsg'
Learn More

p-Chlorophenylalanine, Tocris Bioscience™

Product Code. 18782474
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Tryptophan hydroxylase inhibitor

Binds irreversibly to tryptophan hydroxylase to cause depletion of serotonin in the brain.

Specifications

CAS 7424-00-2
Molecular Formula C9H10ClNO2
MDL Number MFCD00002601
Synonym fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O
IUPAC Name 2-amino-3-(4-chlorophenyl)propanoic acid
Molecular Weight (g/mol) 199.63
PubChem CID 4652
Formula Weight 199.64
Percent Purity >99%
Quantity 100 mg
Chemical Name or Material p-Chlorophenylalanine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'