missing translation for 'onlineSavingsMsg'
Learn More

p-Coumaric Acid, 98+%, (Crystalline), MP Biomedicals

Product Code. 11468200 Shop All MP Biomedicals Products
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Predominantly trans-isomer

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7400-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key NGSWKAQJJWESNS-ZZXKWVIFSA-N
Synonym p-coumaric acid, 4-hydroxycinnamic acid, p-hydroxycinnamic acid, 4-coumaric acid, trans-4-hydroxycinnamic acid, trans-p-coumaric acid, p-cumaric acid, 3-4-hydroxyphenyl acrylic acid, para-coumaric acid, hydroxycinnamic acid
PubChem CID 637542
ChEBI CHEBI:32374
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)O

Specifications

Assay Percent Range 98+%
Quantity 10 g
Formula Weight 164.2
Physical Form Crystalline
Chemical Name or Material p-Coumaric Acid
compliance-icons
Product Identifier
  • p-Coumaric Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 4-hydroxycinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Coumaric Acid, 98+%, (Crystalline), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.